This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299379_m1
See other TRAPPC13 GE Assays ›
Gene Symbol:
Trappc13
Gene Name:
trafficking protein particle complex 13
Gene Aliases:
2410002O22Rik, 2610524F24Rik, AI451892, AU017229
Chromosome Location:
Chr.13: 104142153 - 104178703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299379_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001093759.1 NP_001087228.1 5 - 6 730 91
NM_001093760.1 NP_001087229.1 5 - 6 730 91
NM_025879.2 NP_080155.3 5 - 6 730 91
GenBank mRNA AK010346.1 - 5 - 6 711 91
AK013956.1 - 5 - 6 688 91
AK084373.1 - 5 - 6 680 91
AK149832.1 - 5 - 6 733 91
AK163342.1 - 5 - 6 718 91
AK167749.1 - 5 - 6 690 91
BC021756.1 - 5 - 6 671 3584010 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002596.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_001093759.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_001093759.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_001093760.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_001093760.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_025879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 NM_025879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017676.mm.1 TC1300002596.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_001093759.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_001093759.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_001093760.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_001093760.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_025879.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 NM_025879.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017677.mm.1 TC1300002596.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0