This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299490_m1
See other PLA2G6 GE Assays ›
Gene Symbol:
Pla2g6
Gene Name:
phospholipase A2, group VI
Gene Aliases:
BB112799, PNPLA9, iPLA(2)beta, iPLA2, iPLA2beta
Chromosome Location:
Chr.15: 79286228 - 79328530 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299490_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199023.1 NP_001185952.1 5 - 6 1579 64
NM_001199024.1 NP_001185953.1 5 - 6 1579 64
NM_001199025.1 NP_001185954.1 6 - 7 1084 64
NM_016915.4 NP_058611.1 5 - 6 1125 64
GenBank mRNA AF259401.1 - 4 - 5 859 64
AK144495.1 - 6 - 7 958 64
AK153333.1 - 5 - 6 1385 64
BC003487.1 - 5 - 6 918 2811357 64
BC049778.1 - 5 - 6 1305 64
BC052845.1 - 5 - 6 950 30054573 64
BC057209.1 - 5 - 6 1172 5329147 64
DV647961.1 - 1 - 2 79 64
U88624.2 - 4 - 5 801 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001868.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g6

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
lipid metabolic process
chemotaxis
positive regulation of cytosolic calcium ion concentration
memory
metabolic process
urinary bladder smooth muscle contraction
lipid catabolic process
cardiolipin biosynthetic process
response to endoplasmic reticulum stress
positive regulation of insulin secretion involved in cellular response to glucose stimulus
positive regulation of vasodilation
positive regulation of exocytosis
negative regulation of synaptic transmission, glutamatergic
positive regulation of protein kinase C signaling
positive regulation of release of cytochrome c from mitochondria
positive regulation of arachidonic acid secretion
regulation of store-operated calcium channel activity
positive regulation of ceramide biosynthetic process
phospholipase A2 activity
calmodulin binding
hydrolase activity
protein kinase binding
ATP-dependent protein binding
calcium-independent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199023.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199023.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199024.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199024.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199025.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_001199025.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_016915.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 NM_016915.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500014600.mm.1 TC1500001868.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199023.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199023.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199024.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199024.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199025.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_001199025.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_016915.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 NM_016915.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500014601.mm.1 TC1500001868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0