This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299543_m1
See other HDAC4 GE Assays ›
Gene Symbol:
Hdac4
Gene Name:
histone deacetylase 4
Gene Aliases:
4932408F19Rik, AI047285
Chromosome Location:
Chr.1: 91928779 - 92180385 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299543_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207225.1 NP_997108.1 10 - 11 1401 70
GenBank mRNA AK029933.1 - 7 - 8 938 70
AK155250.1 - 11 - 12 1587 70
BC066052.1 - 10 - 11 1401 6827645 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002806.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac4

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
osteoblast development
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
positive regulation of cell proliferation
negative regulation of cell proliferation
positive regulation of lamellipodium assembly
negative regulation of myotube differentiation
regulation of cardiac muscle contraction by calcium ion signaling
response to denervation involved in regulation of muscle adaptation
positive regulation of smooth muscle cell migration
chromatin modification
histone deacetylation
positive regulation of protein sumoylation
peptidyl-lysine deacetylation
regulation of gene expression, epigenetic
response to drug
regulation of protein binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of neuron apoptotic process
negative regulation of osteoblast differentiation
negative regulation of glycolytic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of smooth muscle cell proliferation
regulation of skeletal muscle fiber development
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of striated muscle cell differentiation
response to interleukin-1
histone H3 deacetylation
histone H4 deacetylation
positive regulation of reactive oxygen species biosynthetic process
RNA polymerase III transcription factor binding
core promoter binding
DNA binding
chromatin binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
zinc ion binding
hydrolase activity
protein kinase binding
potassium ion binding
histone deacetylase activity (H3-K14 specific)
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
activating transcription factor binding
histone deacetylase activity (H4-K16 specific)
histone deacetylase binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
repressing transcription factor binding
NAD-dependent histone deacetylase activity (H3-K18 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6s775:80/100.66.79.102:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0