This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01299853_m1
See other P3H4 GE Assays ›
Gene Symbol:
P3h4
Gene Name:
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Gene Aliases:
1110036O03Rik, AI413214, Leprel4, Sc65
Chromosome Location:
Chr.11: 100408748 - 100414819 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01299853_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P3h4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_176830.2 NP_789800.1 3 - 4 967 68
GenBank mRNA AK029271.1 - 3 - 4 847 68
BC031856.1 - 3 - 4 967 3968941 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003770.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene P3h4

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100034638.mm.1 NM_176830.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034638.mm.1 NM_176830.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100034638.mm.1 TC1100003770.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline