This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01300193_g1
See other GCAT GE Assays ›
Gene Symbol:
Gcat
Gene Name:
glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase)
Gene Aliases:
AI526977, Kbl
Chromosome Location:
Chr.15: 79030874 - 79042531 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01300193_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161712.1 NP_001155184.1 7 - 8 1034 69
NM_013847.4 NP_038875.3 7 - 8 1034 69
GenBank mRNA AF093403.1 - 7 - 8 992 69
AK007395.1 - 7 - 8 967 69
AK013138.1 - 7 - 8 989 69
AK075782.1 - 7 - 8 974 69
AK076469.1 - 7 - 8 1007 69
AK087433.1 - 7 - 8 993 69
BC024107.1 - 7 - 8 1034 5125361 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000663.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcat

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363920 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363918 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363919 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363921 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK087433.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 BC024107.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK013138.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK075782.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AF093403.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK087433.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 BC024107.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK013138.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AK075782.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159402 AF093403.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652845 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370274 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 774045 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475474 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965407 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161613 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 721 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK087433.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 BC024107.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK013138.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK075782.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AF093403.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK087433.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 BC024107.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK013138.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AK075782.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159402 AF093403.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK087433.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 BC024107.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK013138.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK075782.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AF093403.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK087433.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 BC024107.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK013138.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AK075782.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P159402 AF093403.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363954 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363970 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363959 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363925 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363931 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363968 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363919 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363966 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363917 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363921 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363971 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363928 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363922 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363930 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363958 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363918 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363962 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363924 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363920 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30363964 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078097 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08429196 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03078079 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08429182 - -
Whole Mouse Genome G4122A A_51_P159402 AK087433.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 BC024107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AK013138.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AK075782.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AF093403.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AK087433.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 BC024107.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AK013138.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AK075782.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P159402 AF093403.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109250 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134406 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12133865 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12133871 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12133870 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363918 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363921 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363919 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30363920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363997 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363896 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363971 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364004 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363898 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364024 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363950 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364019 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363990 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363968 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363992 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363976 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363880 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364018 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363962 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364008 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363974 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363952 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363948 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364007 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364001 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363953 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364021 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363989 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363955 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363999 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363869 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363996 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363921 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363959 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364003 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364014 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363977 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363964 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364031 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363942 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363979 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364005 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363986 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363871 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363966 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363972 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363988 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363937 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364006 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364011 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363887 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30363889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30364015 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363944 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12463909 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363934 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363936 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12463886 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363919 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363932 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363924 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363953 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363968 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363950 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363946 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11869882 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363962 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363945 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363964 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363933 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12274379 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363971 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12133894 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363939 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363920 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363952 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363966 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363955 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12133870 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363918 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363937 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363940 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363970 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363958 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363925 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30363959 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10425214 - -
Affymetrix Mouse Gene 1.0 ST Array 10425213 - -
Affymetrix Mouse Gene 1.0 ST Array 10425215 - -
Affymetrix Mouse Gene 1.0 ST Array 10425216 - -
Affymetrix Mouse Gene 1.0 ST Array 10425224 - -
Affymetrix Mouse Gene 1.0 ST Array 10425221 - -
Affymetrix Mouse Gene 1.0 ST Array 10425217 - -
Affymetrix Mouse Gene 1.0 ST Array 10425222 - -
Affymetrix Mouse Gene 1.0 ST Array 10425219 - -
Affymetrix Mouse Gene 1.0 ST Array 10425218 - -
Affymetrix Mouse Gene 1.0 ST Array 10425220 - -
Affymetrix Mouse Gene 1.0 ST Array 10425212 - -
Affymetrix HT MG-430 PM Array Plate 1417824_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1417823_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10425211 - -
Affymetrix Mouse Gene 1.1 ST Array 10425223 - -
Mouse Genome 430 2.0 Array 1417823_PM_at - -
Mouse Genome 430 2.0 Array 1417824_at - -
Mouse Genome 430 2.0 Array 1417823_at - -
Mouse Genome 430 2.0 Array 1417824_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 26912_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 26912_PM_at - -
Affymetrix Mouse Clariom S Array PSR1500004935.mm.1 NM_001161712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004935.mm.1 NM_001161712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004935.mm.1 NM_015738.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004935.mm.1 NM_015738.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500004935.mm.1 TC1500000663.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004932.mm.1 NM_013847.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004932.mm.1 NM_013847.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500004932.mm.1 TC1500000663.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004936.mm.1 NM_001161712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004936.mm.1 NM_001161712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004936.mm.1 TC1500000663.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004931.mm.1 NM_013847.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004931.mm.1 NM_013847.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500004931.mm.1 TC1500000663.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500004930.mm.1 NM_001161712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004930.mm.1 NM_001161712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004930.mm.1 NM_013847.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004930.mm.1 NM_013847.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500004930.mm.1 TC1500000663.mm.1 MTA TC
Mouse Expression Set 430 1417823_PM_at - -
Mouse Expression Set 430 1417823_at - -
Mouse Expression Set 430 1417824_at - -
Mouse Expression Set 430 1417824_PM_at - -
Murine Genome U74v2 Set 160628_at - -
Murine Genome U74v2 Set 160628_PM_at - -
Mouse Genome 430A 2.0 Array 1417823_at - -
Mouse Genome 430A 2.0 Array 1417824_at - -
Mouse Genome 430A 2.0 Array 1417824_PM_at - -
Mouse Genome 430A 2.0 Array 1417823_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417824_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1417824_at - -
Affymetrix HT Mouse Genome 430A Array 1417823_at - -
Affymetrix HT Mouse Genome 430A Array 1417823_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 5270672 - -
MouseWG-6 v2 Expression BeadChip 102470390 - -
Illumina mouse-6 v1.0 expression beadchip 102470390 - -
Illumina mouse-6 v1.0 expression beadchip 5270672 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005270672 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2777696 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1248083 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2777696 - -
mouse-6 v1.1 (Illumina) scl026912.9_26-S - -
Illumina Sentrix mouse-6.v1 bead array 5270672 - -
Illumina Sentrix mouse-6.v1 bead array 102470390 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl026912.9_26-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270672 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 102470390 - -
MouseRef-8 v2 Expression BeadChip ILMN_2875336 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248083 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2875336 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2777696 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl026912.9_26-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0