This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01301056_m1
See other LMF1 GE Assays ›
Gene Symbol:
Lmf1
Gene Name:
lipase maturation factor 1
Gene Aliases:
2400010G15Rik, AW822050, Tmem112, cld
Chromosome Location:
Chr.17: 25579174 - 25662826 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01301056_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lmf1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029624.4 NP_083900.3 1 - 2 208 93
GenBank mRNA AK010312.1 - 1 - 2 179 93
AK035119.1 - 1 - 2 196 93
AK053241.1 - 1 - 2 185 93
AK135504.1 - 1 - 2 184 93
AK142235.1 - 1 - 2 184 93
AK154622.1 - 1 - 2 205 93
AK169983.1 - 1 - 2 206 93
AK172374.1 - 1 - 2 187 93
BC020104.1 - 1 - 2 215 3963699 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000380.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lmf1

Gene Ontology Categories:

Function(s) Process(es)

ER to Golgi vesicle-mediated transport
protein secretion
protein glycosylation in Golgi
chylomicron remnant clearance
positive regulation of lipoprotein lipase activity
protein maturation
regulation of cholesterol metabolic process
regulation of triglyceride metabolic process
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700003148.mm.1 NM_029624.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700003148.mm.1 NM_029624.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700003148.mm.1 TC1700000380.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 742093 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0