This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302051_g1
See other PDK2 GE Assays ›
Gene Symbol:
Pdk2
Gene Name:
pyruvate dehydrogenase kinase, isoenzyme 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 95026258 - 95041371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302051_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pdk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133667.2 NP_598428.2 10 - 11 1199 64
GenBank mRNA AF267660.1 - 10 - 11 1160 64
AK002647.1 - 10 - 11 1143 64
BC021764.1 - 10 - 11 1153 3488813 64
CK618575.1 - 3 - 4 254 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003583.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pdk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glucose metabolic process
regulation of gluconeogenesis
protein phosphorylation
regulation of pH
insulin receptor signaling pathway
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of cellular ketone metabolic process
regulation of glucose metabolic process
phosphorylation
peptidyl-serine phosphorylation
cellular response to nutrient
cellular response to reactive oxygen species
glucose homeostasis
intrinsic apoptotic signaling pathway by p53 class mediator
nucleotide binding
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
kinase activity
transferase activity
protein complex binding
protein homodimerization activity
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036286 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036285 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036287 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011802 AF267660.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4059 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 65787 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536404 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26886 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899521 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23089 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649705 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 315679 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264715 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151370 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139517 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011802 AF267660.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426276 AF267660.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036300 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036295 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036274 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036289 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036278 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036264 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036268 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036291 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036286 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036269 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036293 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036283 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036287 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036272 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036266 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036297 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036281 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036276 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036279 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036271 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036284 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036292 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036299 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28036288 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02400135 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07449391 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139517 - -
Whole Mouse Genome G4122A A_51_P426276 AF267660.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P426276 AF267660.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111183 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126630 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139517 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383667 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383665 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383683 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09383677 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08978324 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036285 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036286 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28036287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036276 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036209 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036203 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036287 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036244 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036300 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036148 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036187 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036281 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036248 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036241 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036292 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036129 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036293 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036266 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036253 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036245 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036259 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036230 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036289 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036288 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036162 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036268 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036213 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036214 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036269 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036130 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036234 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036154 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036225 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036178 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036279 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036122 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036239 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036284 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036256 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28036263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09380289 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036249 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036263 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036253 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036235 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036288 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036241 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09383685 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036246 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036276 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08979463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036260 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036245 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09383665 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036274 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036301 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036259 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036271 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09383673 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08978327 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09383677 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036262 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09349542 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08979465 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036269 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036266 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036295 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036291 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09383683 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09094766 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036230 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036248 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09349541 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036251 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09179616 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P28036234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09349547 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10390116 - -
Affymetrix Mouse Gene 1.0 ST Array 10390104 - -
Affymetrix Mouse Gene 1.0 ST Array 10390113 - -
Affymetrix Mouse Gene 1.0 ST Array 10390107 - -
Affymetrix Mouse Gene 1.0 ST Array 10390106 - -
Affymetrix Mouse Gene 1.0 ST Array 10390112 - -
Affymetrix Mouse Gene 1.0 ST Array 10390115 - -
Affymetrix Mouse Gene 1.0 ST Array 10390114 - -
Affymetrix Mouse Gene 1.0 ST Array 10390110 - -
Affymetrix Mouse Gene 1.0 ST Array 10390108 - -
Affymetrix Mouse Gene 1.0 ST Array 10390111 - -
Affymetrix Mouse Gene 1.0 ST Array 10390109 - -
Affymetrix Mouse Gene 1.0 ST Array 10390105 - -
Affymetrix HT MG-430 PM Array Plate 1448825_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10390103 - -
Mouse Genome 430 2.0 Array 1448825_at - -
Mouse Genome 430 2.0 Array 1448825_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18604_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18604_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100032833.mm.1 NM_133667.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032833.mm.1 NM_133667.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032833.mm.1 TC1100003583.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032834.mm.1 NM_133667.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032834.mm.1 NM_133667.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100032834.mm.1 TC1100003583.mm.1 MTA TC
Mouse Expression Set 430 1448825_at - -
Mouse Expression Set 430 1448825_PM_at - -
Mouse Genome 430A 2.0 Array 1448825_PM_at - -
Mouse Genome 430A 2.0 Array 1448825_at - -
Affymetrix HT Mouse Genome 430A Array 1448825_at - -
Affymetrix HT Mouse Genome 430A Array 1448825_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 2690017 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002690017 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737658 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2737658 - -
mouse-6 v1.1 (Illumina) scl39696.12_14-S - -
Illumina Sentrix mouse-6.v1 bead array 2690017 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl39696.12_14-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2690017 - -
MouseRef-8 v2 Expression BeadChip ILMN_2737658 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737658 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl39696.12_14-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0