This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302304_g1
See other HAP1 GE Assays ›
Gene Symbol:
Hap1
Gene Name:
huntingtin-associated protein 1
Gene Aliases:
HAP-1
Chromosome Location:
Chr.11: 100347327 - 100356353 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302304_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010404.3 NP_034534.1 7 - 8 83
NM_177981.2 NP_817090.1 7 - 8 83
GenBank mRNA AJ000262.1 - 7 - 8 1072 83
AJ002271.1 - 7 - 8 1072 83
BC034089.1 - 7 - 8 4481735 83
BC053043.1 - 7 - 8 5694628 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003766.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hap1

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
exocytosis
autophagy
anterograde axonal transport
retrograde axonal transport
protein localization
positive regulation of neuron projection development
protein transport
regulation of exocytosis
cerebellum development
hypothalamus cell differentiation
cell projection organization
neuron projection development
positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
positive regulation of synaptic transmission, GABAergic
positive regulation of neurotrophin production
positive regulation of epidermal growth factor receptor signaling pathway
vesicle transport along microtubule
neurotrophin TRK receptor signaling pathway
positive regulation of neurogenesis
positive regulation of inclusion body assembly
negative regulation of beta-amyloid formation
regulation of organelle transport along microtubule
positive regulation of nonmotile primary cilium assembly
receptor binding
protein binding
protein domain specific binding
identical protein binding
ion channel binding
brain-derived neurotrophic factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100034571.mm.1 NM_010404.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034571.mm.1 NM_010404.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100034571.mm.1 NM_177981.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034571.mm.1 NM_177981.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100034571.mm.1 TC1100003766.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline