This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302369_g1
See other TRAPPC6A GE Assays ›
Gene Symbol:
Trappc6a
Gene Name:
trafficking protein particle complex 6A
Gene Aliases:
1810073E21Rik, 4930519D19Rik, AI480686, TRS33, mhyp
Chromosome Location:
Chr.7: 19507406 - 19516145 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302369_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trappc6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025960.3 NP_080236.2 2 - 3 171 158
GenBank mRNA AK007965.1 - 2 - 3 170 158
AK008022.1 - 2 - 3 171 158
AK008462.2 - 2 - 3 170 158
AK088832.1 - 2 - 3 158
BC037154.1 - 2 - 3 208 2811950 158
BC106114.1 - 2 - 3 185 4168174 158

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000303.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trappc6a

Gene Ontology Categories:

Function(s)

transport
vesicle-mediated transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700002425.mm.1 TC0700000303.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700002425.mm.1 NM_025960.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0