This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302422_m1
See other MMP28 GE Assays ›
Gene Symbol:
Mmp28
Gene Name:
matrix metallopeptidase 28 (epilysin)
Gene Aliases:
D130023P12
Chromosome Location:
Chr.11: 83441241 - 83463064 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302422_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmp28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080453.2 NP_536701.1 1 - 2 219 143
NM_172797.2 NP_766385.1 1 - 2 219 143
GenBank mRNA AK051258.1 - 1 - 2 282 143
AY065653.1 - 1 - 2 219 143
AY071828.1 - 1 - 2 219 143
AY071829.1 - 1 - 2 219 143
BC137921.1 - 1 - 2 174 8860940 143
BC145048.1 - 1 - 2 174 9053596 143
BC145049.1 - 1 - 2 174 9053597 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003394.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmp28

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
negative regulation of macrophage chemotaxis
metalloendopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100031093.mm.1 NM_080453.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031093.mm.1 NM_172797.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031093.mm.1 TC1100003394.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100031095.mm.1 NM_080453.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031095.mm.1 NM_172797.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031095.mm.1 TC1100003394.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100031096.mm.1 NM_080453.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031096.mm.1 NM_172797.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031096.mm.1 TC1100003394.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0