This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302447_m1
See other TADA2A GE Assays ›
Gene Symbol:
Tada2a
Gene Name:
transcriptional adaptor 2A
Gene Aliases:
AV319371, D030022J10Rik, Tada2l
Chromosome Location:
Chr.11: 84078920 - 84129642 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302447_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tada2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172562.3 NP_766150.1 11 - 12 937 64
GenBank mRNA AK157938.1 - 11 - 12 930 64
AK177030.1 - 1 - 2 99 64
AK209681.1 - 1 - 2 99 64
AK216505.1 - 3 - 4 196 64
BC005610.1 - 8 - 9 664 64
BC025448.1 - 6 - 7 429 64
BC038821.1 - 11 - 12 940 3990023 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003407.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tada2a

Panther Classification:

Molecular Function -

DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
mitotic nuclear division
histone acetylation
regulation of histone deacetylation
regulation of protein stability
histone H3 acetylation
regulation of tubulin deacetylation
DNA binding
chromatin binding
histone acetyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100031186.mm.1 NM_172562.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100031186.mm.1 NM_172562.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031186.mm.1 TC1100003407.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100031187.mm.1 NM_172562.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100031187.mm.1 NM_172562.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100031187.mm.1 TC1100003407.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0