This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302468_g1
See other PRSS33 GE Assays ›
Gene Symbol:
Prss33
Gene Name:
protease, serine 33
Gene Aliases:
Eos, mT6, tryptase-6
Chromosome Location:
Chr.17: 23833338 - 23836126 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302468_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081399.2 NP_001074868.2 4 - 5 546 86
GenBank mRNA AY262280.1 - 4 - 5 522 86
BC101950.1 - 4 - 5 522 40044230 86
BC103541.1 - 5 - 6 618 40044224 86
BC103542.1 - 4 - 5 522 40044219 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001607.mm.1

Set Membership:

Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700014080.mm.1 NM_001081399.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700014080.mm.1 NM_001081399.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700014080.mm.1 TC1700001607.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700014081.mm.1 NM_001081399.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700014081.mm.1 NM_001081399.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700014081.mm.1 TC1700001607.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0