This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302514_m1
See other FGF13 GE Assays ›
Gene Symbol:
Fgf13
Gene Name:
fibroblast growth factor 13
Gene Aliases:
Fhf2
Chromosome Location:
Chr.X: 59062139 - 59585572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302514_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290415.1 NP_001277344.1 1 - 2 78 108
GenBank mRNA AF199614.1 - 1 - 2 162 108
AK141848.1 - 1 - 2 78 108
CJ064772.1 - 1 - 2 79 108

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF199614.1, AK141848.1, NM_001290415.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
neuron migration
apoptotic process
negative regulation of microtubule depolymerization
nervous system development
learning
memory
cell proliferation
hippocampus development
cerebral cortex cell migration
establishment of neuroblast polarity
positive regulation of protein kinase activity
microtubule polymerization
negative regulation of collateral sprouting
protein localization to plasma membrane
receptor binding
microtubule binding
growth factor activity
sodium channel regulator activity
protein kinase activator activity
ion channel binding
beta-tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline