This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302515_m1
See other FGF13 GE Assays ›
Gene Symbol:
Fgf13
Gene Name:
fibroblast growth factor 13
Gene Aliases:
Fhf2
Chromosome Location:
Chr.X: 59062139 - 59585572 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302515_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fgf13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290414.1 NP_001277343.1 4 - 5 676 70
NM_001290415.1 NP_001277344.1 2 - 3 187 70
NM_010200.3 NP_034330.2 2 - 3 671 70
GenBank mRNA AF020737.1 - 2 - 3 304 70
AK141848.1 - 2 - 3 187 70
BC018238.1 - 2 - 3 671 4481123 70
CJ064772.1 - 2 - 3 188 70
U66202.1 - 2 - 3 298 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002292.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fgf13

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
neuron migration
apoptotic process
negative regulation of microtubule depolymerization
nervous system development
learning
memory
cell proliferation
hippocampus development
cerebral cortex cell migration
establishment of neuroblast polarity
positive regulation of protein kinase activity
microtubule polymerization
negative regulation of collateral sprouting
protein localization to plasma membrane
receptor binding
microtubule binding
growth factor activity
sodium channel regulator activity
protein kinase activator activity
ion channel binding
beta-tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00014486.mm.1 NM_010200.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00014486.mm.1 TC0X00002292.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00014485.mm.1 NM_010200.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00014485.mm.1 TC0X00002292.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline