This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302623_m1
See other HAGHL GE Assays ›
Gene Symbol:
Haghl
Gene Name:
hydroxyacylglutathione hydrolase-like
Gene Aliases:
1500017E18Rik, 2810014I23Rik, C330022E15Rik
Chromosome Location:
Chr.17: 25779843 - 25785669 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302623_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Haghl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271436.1 NP_001258365.1 1 - 2 164 59
GenBank mRNA AK021220.1 - 1 - 2 161 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001271436.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

glutathione biosynthetic process
biological_process
molecular_function
hydroxyacylglutathione hydrolase activity
zinc ion binding
CoA hydrolase activity
hydrolase activity
2-oxoglutaryl-CoA thioesterase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
3-isopropylbut-3-enoyl-CoA thioesterase activity
glutaryl-CoA hydrolase activity
metal ion binding
acyl-CoA hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline