| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_001252625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_001252625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_001252626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_001252626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_029880.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
NM_029880.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005753.mm.1 |
TC1200000806.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_001252625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_001252625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_001252626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_001252626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_029880.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
NM_029880.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005767.mm.1 |
TC1200000806.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_001252625.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_001252625.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_001252626.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_001252626.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_029880.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
NM_029880.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200005763.mm.1 |
TC1200000806.mm.1 |
MTA TC |