This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01302818_m1
See other PTGR2 GE Assays ›
Gene Symbol:
Ptgr2
Gene Name:
prostaglandin reductase 2
Gene Aliases:
1810016I24Rik, 9130222H03Rik, 9630002F03Rik, AI838763, B830026H24Rik, PGR-2, PRG-2, Zadh1
Chromosome Location:
Chr.12: 84285237 - 84315832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01302818_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptgr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252626.1 NP_001239555.1 2 - 3 265 131
NM_029880.3 NP_084156.2 2 - 3 265 131
GenBank mRNA AK007522.1 - 2 - 3 148 131
AK020666.1 - 2 - 3 194 131
AK036168.1 - 2 - 3 187 131
AK040396.1 - 2 - 3 197 131
AK159932.1 - 2 - 3 162 131
AK168895.1 - 2 - 3 156 131
BC021466.1 - 2 - 3 211 5068419 131
CJ050177.1 - 2 - 3 266 131
CJ137771.1 - 2 - 3 330 131

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK036168.1, AK159932.1, AK168895.1, BC021466.1, NM_001252626.1, NM_029880.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptgr2

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin metabolic process
oxidation-reduction process
zinc ion binding
oxidoreductase activity
13-prostaglandin reductase activity
15-oxoprostaglandin 13-oxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline