This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303097_m1
See other LIG3 GE Assays ›
Gene Symbol:
Lig3
Gene Name:
ligase III, DNA, ATP-dependent
Gene Aliases:
D11Wsu78e
Chromosome Location:
Chr.11: 82781109 - 82804274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303097_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lig3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291245.1 NP_001278174.1 11 - 12 1997 138
NM_001291246.1 NP_001278175.1 11 - 12 1994 138
NM_001291247.1 NP_001278176.1 11 - 12 1998 138
NM_010716.3 NP_034846.2 11 - 12 2001 138
GenBank mRNA AK031540.1 - 11 - 12 1938 138
AK143531.1 - 11 - 12 1867 138
AK150644.1 - 11 - 12 1906 138
AK156018.1 - 11 - 12 1916 138
BC026143.1 - 11 - 12 1969 138
BC049240.1 - 11 - 12 1904 4504384 138
BC083500.1 - 11 - 12 1900 3673103 138

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001232.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lig3

Panther Classification:

Molecular Function -

DNA ligase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA ligation
DNA repair
base-excision repair, DNA ligation
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
mitochondrion organization
cell cycle
mitochondrial DNA repair
negative regulation of DNA recombination
DNA ligation involved in DNA repair
cell division
DNA biosynthetic process
negative regulation of mitochondrial DNA replication
double-strand break repair via alternative nonhomologous end joining
nucleotide binding
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
zinc ion binding
ligase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100011533.mm.1 NM_010716.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100011533.mm.1 TC1100001232.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100011536.mm.1 NM_010716.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100011536.mm.1 TC1100001232.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-k52cq:80/100.66.77.40:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0