This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303098_g1
See other LIG3 GE Assays ›
Gene Symbol:
Lig3
Gene Name:
ligase III, DNA, ATP-dependent
Gene Aliases:
D11Wsu78e
Chromosome Location:
Chr.11: 82781109 - 82804274 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303098_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lig3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291245.1 NP_001278174.1 12 - 13 2084 79
NM_001291246.1 NP_001278175.1 12 - 13 2081 79
NM_001291247.1 NP_001278176.1 12 - 13 2085 79
NM_010716.3 NP_034846.2 12 - 13 2088 79
GenBank mRNA AK031540.1 - 12 - 13 2025 79
AK143531.1 - 12 - 13 1954 79
AK150644.1 - 12 - 13 1993 79
AK156018.1 - 12 - 13 2003 79
BC049240.1 - 12 - 13 1991 4504384 79
BC083500.1 - 12 - 13 1987 3673103 79
U66057.1 - 12 - 13 2019 79
U66058.1 - 12 - 13 2019 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lig3

Panther Classification:

Molecular Function -

DNA ligase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA ligation
DNA repair
base-excision repair, DNA ligation
double-strand break repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
mitochondrion organization
cell cycle
mitochondrial DNA repair
negative regulation of DNA recombination
DNA ligation involved in DNA repair
cell division
DNA biosynthetic process
negative regulation of mitochondrial DNA replication
double-strand break repair via alternative nonhomologous end joining
nucleotide binding
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
protein binding
ATP binding
zinc ion binding
ligase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0