This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303303_m1
See other RACGAP1 GE Assays ›
Gene Symbol:
Racgap1
Gene Name:
Rac GTPase-activating protein 1
Gene Aliases:
AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
Chromosome Location:
Chr.15: 99620496 - 99651656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303303_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Racgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253808.1 NP_001240737.1 9 - 10 1038 78
NM_001253809.1 NP_001240738.1 9 - 10 1034 78
NM_012025.7 NP_036155.1 10 - 11 1169 78
GenBank mRNA AB030252.1 - 9 - 10 1018 78
AF079974.1 - 9 - 10 979 78
AF212320.1 - 9 - 10 1005 78
AF212321.1 - 10 - 11 1136 78
AK053850.1 - 9 - 10 1000 78
AK144608.1 - 10 - 11 1137 78
AK144745.1 - 9 - 10 998 78
AK154929.1 - 10 - 11 1157 78
AK162660.1 - 7 - 8 4238 78
AK165897.1 - 9 - 10 991 78
AK166084.1 - 10 - 11 1127 78
AK166094.1 - 9 - 10 996 78
AK168020.1 - 9 - 10 995 78
AK168025.1 - 9 - 10 995 78
AK168170.1 - 9 - 10 995 78
AK168679.1 - 9 - 10 979 78
BC010715.1 - 9 - 10 984 3602242 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002217.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Racgap1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
actomyosin contractile ring assembly
transport
ion transport
cell cycle
signal transduction
multicellular organism development
spermatogenesis
sulfate transport
embryo development
cell differentiation
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
mitotic spindle midzone assembly
cell division
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
lipid binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018165.mm.1 TC1500002217.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0