This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303309_m1
See other RACGAP1 GE Assays ›
Gene Symbol:
Racgap1
Gene Name:
Rac GTPase-activating protein 1
Gene Aliases:
AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
Chromosome Location:
Chr.15: 99620496 - 99651656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303309_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Racgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253808.1 NP_001240737.1 2 - 3 241 76
NM_001253809.1 NP_001240738.1 2 - 3 237 76
NM_012025.7 NP_036155.1 3 - 4 372 76
GenBank mRNA AB030252.1 - 2 - 3 221 76
AF079974.1 - 2 - 3 182 76
AF212320.1 - 2 - 3 208 76
AF212321.1 - 3 - 4 339 76
AK053850.1 - 2 - 3 202 76
AK144608.1 - 3 - 4 340 76
AK144745.1 - 2 - 3 200 76
AK154929.1 - 3 - 4 360 76
AK162660.1 - 2 - 3 168 76
AK165897.1 - 2 - 3 194 76
AK166094.1 - 2 - 3 199 76
AK168020.1 - 2 - 3 198 76
AK168025.1 - 2 - 3 198 76
AK168170.1 - 2 - 3 198 76
AK168679.1 - 2 - 3 182 76
BC010715.1 - 2 - 3 187 3602242 76
BY309801.1 - 2 - 3 241 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Racgap1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
actomyosin contractile ring assembly
transport
ion transport
cell cycle
signal transduction
multicellular organism development
spermatogenesis
sulfate transport
embryo development
cell differentiation
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
mitotic spindle midzone assembly
cell division
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
lipid binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0