This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303639_m1
See other GHR GE Assays ›
Gene Symbol:
Ghr
Gene Name:
growth hormone receptor
Gene Aliases:
GHBP, GHR/BP
Chromosome Location:
Chr.15: 3317755 - 3583527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303639_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ghr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001048178.2 NP_001041643.1 3 - 4 494 71
NM_001286370.1 NP_001273299.1 3 - 4 195 71
NM_010284.3 NP_034414.2 3 - 4 494 71
GenBank mRNA AK053579.1 - 3 - 4 355 71
AK143886.1 - 3 - 4 196 71
AK143925.1 - 3 - 4 292 71
BC024375.1 - 3 - 4 329 71
BC056645.1 - 3 - 4 310 71
M31680.1 - 3 - 4 230 71
M33324.1 - 3 - 4 230 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001136.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ghr

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
allantoin metabolic process
citrate metabolic process
2-oxoglutarate metabolic process
succinate metabolic process
oxaloacetate metabolic process
isoleucine metabolic process
valine metabolic process
creatine metabolic process
fatty acid metabolic process
endocytosis
JAK-STAT cascade
hormone-mediated signaling pathway
cytokine-mediated signaling pathway
taurine metabolic process
response to estradiol
cellular response to hormone stimulus
regulation of multicellular organism growth
positive regulation of multicellular organism growth
activation of Janus kinase activity
activation of JAK2 kinase activity
multicellular organism metabolic process
positive regulation of JAK-STAT cascade
creatinine metabolic process
insulin-like growth factor receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
growth hormone receptor signaling pathway
negative regulation of neuron death
cytokine receptor activity
growth hormone receptor activity
protein binding
peptide hormone binding
growth factor binding
protein kinase binding
protein phosphatase binding
SH2 domain binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500009179.mm.1 NM_001048178.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500009179.mm.1 NM_001286370.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009179.mm.1 NM_010284.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500009179.mm.1 TC1500001136.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline