This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01303665_g1
See other SKP2 GE Assays ›
Gene Symbol:
Skp2
Gene Name:
S-phase kinase-associated protein 2 (p45)
Gene Aliases:
4930500A04Rik, FBXL1, FWD1, p45
Chromosome Location:
Chr.15: 9111982 - 9155425 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01303665_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Skp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285980.1 NP_001272909.1 6 - 7 957 59
NM_013787.3 NP_038815.1 6 - 7 1027 59
GenBank mRNA AF083215.1 - 6 - 7 890 59
AK006221.1 - 6 - 7 957 59
AK032979.1 - 6 - 7 938 59
AK044212.1 - 6 - 7 928 59
AK166034.1 - 6 - 7 928 59
AK167677.1 - 6 - 7 925 59
BC092236.1 - 6 - 7 901 30610417 59
BC098371.1 - 6 - 7 1010 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001192.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Skp2

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
ubiquitin-dependent protein catabolic process
positive regulation of intracellular estrogen receptor signaling pathway
regulation of apoptotic process
positive regulation of smooth muscle cell proliferation
regulation of cell cycle
positive regulation of protein polyubiquitination
protein binding
identical protein binding
ubiquitin protein ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500009570.mm.1 NM_001285980.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009570.mm.1 NM_013787.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500009570.mm.1 TC1500001192.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0