Assay Details
| Gene Symbol: Fyb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278269.1 | NP_001265198.1 | 11 - 12 | 2005 | 66 | |
| GenBank mRNA | AF001863.1 | - | 11 - 12 | 1981 | 66 | |
| AK038225.1 | - | 11 - 12 | 2033 | 66 | ||
| AK137907.1 | - | 11 - 12 | 1961 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fyb
Genomic Map