This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304076_m1
See other UGT3A2 GE Assays ›
Gene Symbol:
Ugt3a2
Gene Name:
UDP glycosyltransferases 3 family, polypeptide A2
Gene Aliases:
AI313915
Chromosome Location:
Chr.15: 9335551 - 9370955 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304076_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ugt3a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144845.3 NP_659094.1 2 - 3 279 95
GenBank mRNA AK041045.1 - 2 - 3 264 95
AK050128.1 - 2 - 3 280 95
AK052644.1 - 2 - 3 289 95
AK143815.1 - 2 - 3 294 95
BC022134.1 - 2 - 3 264 5098754 95
BC034837.1 - 2 - 3 260 5134131 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000073.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ugt3a2

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
flavonoid biosynthetic process
cellular response to hormone stimulus
flavonoid glucuronidation
cellular response to genistein
UDP-glycosyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500000665.mm.1 NM_144845.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500000665.mm.1 NM_144845.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500000665.mm.1 TC1500000073.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4ln64:80/100.66.79.201:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0