This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304216_m1
See other ASIC2 GE Assays ›
Gene Symbol:
Asic2
Gene Name:
acid-sensing (proton-gated) ion channel 2
Gene Aliases:
ACIC2, Accn1, BNC1, BNaC1, BNaC1a, Mdeg
Chromosome Location:
Chr.11: 80880163 - 81968396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304216_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001034013.2 NP_001029185.1 6 - 7 1411 119
NM_007384.3 NP_031410.1 6 - 7 1790 119
GenBank mRNA AF348465.1 - 6 - 7 1424 119
AK134778.1 - 6 - 7 1424 119
BC138529.1 - 6 - 7 1641 8861549 119
BC138530.1 - 6 - 7 1639 8861550 119
U57353.1 - 3 - 4 350 119
Y14634.1 - 6 - 7 1790 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003353.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic2

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of systemic arterial blood pressure by aortic arch baroreceptor feedback
transport
ion transport
cation transport
sodium ion transport
phototransduction
sensory perception of sound
response to mechanical stimulus
response to acidic pH
regulation of gene expression
regulation of vasoconstriction
regulation of blood coagulation
ion transmembrane transport
regulation of ion transmembrane transport
protein localization to synapse
cellular response to drug
sodium ion transmembrane transport
regulation of membrane potential
negative regulation of apoptotic process
sensory perception of sour taste
detection of mechanical stimulus involved in sensory perception
positive regulation of synapse assembly
cellular response to acidic pH
cation transmembrane transport
positive regulation of cation channel activity
ion channel activity
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
protein binding
ligand-gated sodium channel activity
ion gated channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline