This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304231_mH
See other MSI2 GE Assays ›
Gene Symbol:
Msi2
Gene Name:
musashi RNA-binding protein 2
Gene Aliases:
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h
Chromosome Location:
Chr.11: 88339382 - 88718267 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304231_mH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msi2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001201341.1 NP_001188270.1 3 - 4 362 59
NM_054043.3 NP_473384.1 3 - 4 324 59
GenBank mRNA AK049637.1 - 3 - 4 324 59
AK049688.1 - 3 - 4 324 59
AK051269.1 - 3 - 4 174 59
AK153928.1 - 3 - 4 197 59
AK156439.1 - 3 - 4 310 59
AK169846.1 - 3 - 4 197 59
BC109368.1 - 3 - 4 277 5249862 59
BC111809.1 - 4 - 5 257 40045350 59

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Msi2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

stem cell development
nucleotide binding
nucleic acid binding
RNA binding
single-stranded RNA binding
poly(U) RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100032055.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032055.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032055.mm.1 NM_054043.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032055.mm.1 NM_054043.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032055.mm.1 TC1100003496.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032050.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032050.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032050.mm.1 NM_054043.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032050.mm.1 NM_054043.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032050.mm.1 TC1100003496.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032051.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032051.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032051.mm.1 NM_054043.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032051.mm.1 NM_054043.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032051.mm.1 TC1100003496.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032048.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032048.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032048.mm.1 NM_054043.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032048.mm.1 NM_054043.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032048.mm.1 TC1100003496.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032049.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032049.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032049.mm.1 NM_054043.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032049.mm.1 NM_054043.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100032049.mm.1 TC1100003496.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100032059.mm.1 NM_001201341.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032059.mm.1 NM_001201341.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100032059.mm.1 TC1100003496.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 736333 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0