This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304422_g1
See other IRAK4 GE Assays ›
Gene Symbol:
Irak4
Gene Name:
interleukin-1 receptor-associated kinase 4
Gene Aliases:
8430405M07Rik, 9330209D03Rik, IRAK-4, NY-REN-64
Chromosome Location:
Chr.15: 94543611 - 94568317 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304422_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Irak4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029926.5 NP_084202.2 4 - 5 665 77
GenBank mRNA AF445803.1 - 4 - 5 653 77
AK020397.2 - 4 - 5 673 77
AK028837.1 - 4 - 5 606 77
AK029028.1 - 4 - 5 666 77
AK139992.1 - 654 77
BC051676.1 - 4 - 5 658 30017484 77

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Irak4

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
immune system process
neutrophil mediated immunity
protein phosphorylation
signal transduction
JNK cascade
phosphorylation
cytokine-mediated signaling pathway
intracellular signal transduction
positive regulation of I-kappaB kinase/NF-kappaB signaling
innate immune response
positive regulation of smooth muscle cell proliferation
neutrophil migration
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
interleukin-1 receptor binding
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0