This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304634_m1
See other SDHC GE Assays ›
Gene Symbol:
Sdhc
Gene Name:
succinate dehydrogenase complex, subunit C, integral membrane protein
Gene Aliases:
0610010E03Rik, AI316496, AU019277
Chromosome Location:
Chr.1: 171129157 - 171150603 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304634_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sdhc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025321.3 NP_079597.2 4 - 5 264 59
GenBank mRNA AK002459.1 - 4 - 5 266 59
AK012818.1 - 4 - 5 265 59
AK032458.1 - 266 59
AK135503.1 - 4 - 5 266 59
BC005779.1 - 4 - 5 251 2810852 59
BC083091.1 - 4 - 5 257 30445339 59
BY711932.1 - 4 - 5 265 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003528.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sdhc

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
mitochondrial electron transport, succinate to ubiquinone
transport
aerobic respiration
oxidation-reduction process
succinate dehydrogenase activity
succinate dehydrogenase (ubiquinone) activity
electron carrier activity
oxidoreductase activity, acting on the CH-CH group of donors
heme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100028860.mm.1 NM_025321.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100028860.mm.1 NM_025321.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100028860.mm.1 TC0100003528.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0