This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304887_m1
See other PTGR2 GE Assays ›
Gene Symbol:
Ptgr2
Gene Name:
prostaglandin reductase 2
Gene Aliases:
1810016I24Rik, 9130222H03Rik, 9630002F03Rik, AI838763, B830026H24Rik, PGR-2, PRG-2, Zadh1
Chromosome Location:
Chr.12: 84285237 - 84315832 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304887_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptgr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252626.1 NP_001239555.1 3 - 4 390 178
GenBank mRNA AK007522.1 - 3 - 4 273 178
AK040396.1 - 3 - 4 322 178

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001252626.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin metabolic process
oxidation-reduction process
zinc ion binding
oxidoreductase activity
13-prostaglandin reductase activity
15-oxoprostaglandin 13-oxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200005769.mm.1 NM_001252626.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005769.mm.1 NM_001252626.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200005769.mm.1 TC1200000806.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline