This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304937_m1
See other TOM1L1 GE Assays ›
Gene Symbol:
Tom1l1
Gene Name:
target of myb1-like 1 (chicken)
Gene Aliases:
2310045L10Rik, C80573, Srcasm
Chromosome Location:
Chr.11: 90645690 - 90687661 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304937_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028011.2 NP_082287.1 8 - 9 897 95
GenBank mRNA AF395837.1 - 8 - 9 861 95
AJ505007.1 - 8 - 9 858 95
AK009826.1 - 6 - 7 632 95
AK033712.1 - 8 - 9 899 95
AK083630.1 - 859 95
BC004710.1 - 1 - 2 1254 3499918 95

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
protein transport
positive regulation of protein autophosphorylation
activation of protein kinase activity
negative regulation of mitotic nuclear division
positive regulation of protein kinase activity
protein binding
SH3 domain binding
protein kinase binding
clathrin binding
protein kinase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0