This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01304960_g1
See other PPP1R1B GE Assays ›
Gene Symbol:
Ppp1r1b
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 1B
Gene Aliases:
AU040756, DARPP-32, Darpp32
Chromosome Location:
Chr.11: 98348738 - 98357795 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01304960_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144828.1 NP_659077.1 2 - 3 498 104
GenBank mRNA AB221544.1 - 2 - 3 147 104
AY640620.1 - 2 - 3 176 104
AY640621.1 - 1 - 2 634 104
BC011122.1 - 2 - 3 408 4164563 104
BC026568.1 - 2 - 3 498 4982294 104
BC031129.1 - 2 - 3 246 4988284 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AY640620.1, AY640621.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r1b

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

response to amphetamine
transcription, DNA-templated
negative regulation of protein kinase activity
signal transduction
negative regulation of female receptivity
visual learning
intracellular signal transduction
negative regulation of catalytic activity
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity
D1 dopamine receptor binding
D2 dopamine receptor binding
D3 dopamine receptor binding
D4 dopamine receptor binding
D5 dopamine receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0