| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100013695.mm.1 |
NM_001284397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013695.mm.1 |
TC1100001474.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_001038664.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_001038664.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_001284393.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_001284397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_023121.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
NM_023121.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013701.mm.1 |
TC1100001474.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_001038664.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_001038664.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_001284393.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_001284397.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_023121.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
NM_023121.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100013700.mm.1 |
TC1100001474.mm.1 |
MTA TC |