Assay Details
| Gene Symbol: Thsd7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172485.3 | NP_766073.2 | 10 - 11 | 2597 | 97 | |
| GenBank mRNA | AK051714.1 | - | 10 - 11 | 2439 | 97 | |
| AK083798.1 | - | 10 - 11 | 2425 | 97 | ||
| BC063250.1 | - | 10 - 11 | 2427 | 6410178 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Thsd7b
Genomic Map