This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305017_m1
See other EML2 GE Assays ›
Gene Symbol:
Eml2
Gene Name:
echinoderm microtubule associated protein like 2
Gene Aliases:
1600029N02Rik
Chromosome Location:
Chr.7: 19176266 - 19206482 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305017_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eml2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162996.1 NP_001156468.1 11 - 12 1152 80
NM_028153.1 NP_082429.1 11 - 12 1209 80
GenBank mRNA AK005562.1 - 11 - 12 1175 80
BC055476.1 - 11 - 12 1152 4218133 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of microtubule nucleation
negative regulation of microtubule polymerization
receptor binding
microtubule binding
protein C-terminus binding
tubulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0