This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305941_m1
See other RACGAP1 GE Assays ›
Gene Symbol:
Racgap1
Gene Name:
Rac GTPase-activating protein 1
Gene Aliases:
AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
Chromosome Location:
Chr.15: 99620496 - 99651656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305941_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Racgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253808.1 NP_001240737.1 1 - 2 151 107
GenBank mRNA AB030252.1 - 1 - 2 131 107
AF079974.1 - 1 - 2 92 105
AF212320.1 - 1 - 2 118 107
AK053850.1 - 1 - 2 112 107
AK144745.1 - 1 - 2 110 107
AK162660.1 - 1 - 2 78 107
AK166094.1 - 1 - 2 108 107
AK168020.1 - 1 - 2 108 107
AK168025.1 - 1 - 2 108 107
AK168170.1 - 1 - 2 108 107
AK168679.1 - 1 - 2 92 107
BC010715.1 - 1 - 2 97 3602242 107
BY309801.1 - 1 - 2 151 107

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB030252.1, AF079974.1, AF212320.1, AK168020.1, AK168025.1, AK168170.1, AK168679.1, BC010715.1, NM_001253808.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
actomyosin contractile ring assembly
transport
ion transport
cell cycle
signal transduction
multicellular organism development
spermatogenesis
sulfate transport
embryo development
cell differentiation
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
mitotic spindle midzone assembly
cell division
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
lipid binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018187.mm.1 TC1500002217.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018191.mm.1 TC1500002217.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018200.mm.1 TC1500002217.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018195.mm.1 TC1500002217.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 443685 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0