This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305949_m1
See other LIMA1 GE Assays ›
Gene Symbol:
Lima1
Gene Name:
LIM domain and actin binding 1
Gene Aliases:
Eplin
Chromosome Location:
Chr.15: 99778468 - 99875456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305949_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lima1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001113545.1 NP_001107017.1 3 - 4 322 73
GenBank mRNA AF307845.1 - 3 - 4 276 73
AK049350.1 - 3 - 4 272 73
BC031490.1 - 3 - 4 248 3498138 73
CJ073082.1 - 3 - 4 323 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002223.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of actin filament depolymerization
ruffle organization
actin filament bundle assembly
actin binding
actin monomer binding
protein binding
zinc ion binding
metal ion binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018249.mm.1 NM_001113545.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018249.mm.1 NM_001113545.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018249.mm.1 NM_023063.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500018249.mm.1 TC1500002223.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0