This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01305998_mH
See other ASIC1 GE Assays ›
Gene Symbol:
Asic1
Gene Name:
acid-sensing (proton-gated) ion channel 1
Gene Aliases:
AI843610, ASIC, ASIC1a, Accn2, B530003N02Rik, BNaC2
Chromosome Location:
Chr.15: 99670266 - 99701130 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01305998_mH
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Asic1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289791.1 NP_001276720.1 2 - 3 1419 89
NM_009597.1 NP_033727.1 4 - 5 897 89
GenBank mRNA AB208022.1 - 2 - 3 803 89
BC067025.1 - 4 - 5 897 6850826 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001021.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Asic1

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
sodium ion transport
calcium ion transport
memory
associative learning
response to acidic pH
monovalent inorganic cation transport
ion transmembrane transport
sodium ion transmembrane transport
regulation of membrane potential
negative regulation of neurotransmitter secretion
sensory perception of sour taste
calcium ion transmembrane transport
cellular response to pH
ion channel activity
cation channel activity
sodium channel activity
protein binding
monovalent inorganic cation transmembrane transporter activity
ligand-gated sodium channel activity
ion gated channel activity
acid-sensing ion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008039.mm.1 NM_001289791.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008039.mm.1 TC1500001021.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008044.mm.1 NM_001289791.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008044.mm.1 NM_009597.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008044.mm.1 NM_009597.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008044.mm.1 TC1500001021.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008040.mm.1 NM_001289791.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008040.mm.1 TC1500001021.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008043.mm.1 NM_001289791.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008043.mm.1 NM_009597.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008043.mm.1 NM_009597.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008043.mm.1 TC1500001021.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0