This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306010_g1
See other SMARCD1 GE Assays ›
Gene Symbol:
Smarcd1
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 1
Gene Aliases:
AA407987, Baf60a, D15Kz1
Chromosome Location:
Chr.15: 99702287 - 99713995 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306010_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031842.2 NP_114030.2 7 - 8 905 64
GenBank mRNA AK075611.1 - 3 - 4 242 64
BC026783.1 - 7 - 8 817 4481097 64
BC059921.1 - 7 - 8 875 64
BC079839.1 - 7 - 8 851 6810462 64
M25773.1 - 7 - 8 861 64
U66620.1 - 7 - 8 861 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001022.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarcd1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

nucleosome disassembly
chromatin remodeling
nervous system development
chromatin modification
chromatin-mediated maintenance of transcription
chromatin binding
receptor binding
protein binding
protein complex scaffold

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008063.mm.1 NM_031842.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008063.mm.1 NM_031842.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008063.mm.1 TC1500001022.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500008062.mm.1 NM_031842.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008062.mm.1 NM_031842.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008062.mm.1 TC1500001022.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-sp9vq:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline