This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306484_m1
See other ABCD2 GE Assays ›
Gene Symbol:
Abcd2
Gene Name:
ATP-binding cassette, sub-family D (ALD), member 2
Gene Aliases:
ABC39, ALDL1, ALDR, ALDRP
Chromosome Location:
Chr.15: 91145871 - 91192504 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306484_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011994.2 NP_036124.2 6 - 7 1848 65
GenBank mRNA AK051445.1 - 6 - 7 1822 65
AK082588.1 - 6 - 7 1820 65
AK087653.1 - 6 - 7 1467 65
AK134763.1 - 1 - 2 159 65
AK161027.1 - 6 - 7 1509 65
BC019187.1 - 6 - 7 1792 5027149 65
Z48670.1 - 6 - 7 1852 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002075.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcd2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
fatty acid beta-oxidation
transport
positive regulation of fatty acid beta-oxidation
very long-chain fatty acid catabolic process
transmembrane transport
nucleotide binding
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016760.mm.1 NM_011994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016760.mm.1 NM_011994.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016760.mm.1 TC1500002075.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0