This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01306722_m1
See other IKZF3 GE Assays ›
Gene Symbol:
Ikzf3
Gene Name:
IKAROS family zinc finger 3
Gene Aliases:
5830411O07Rik, Aiolos, Zfpn1a3, Znfn1a3
Chromosome Location:
Chr.11: 98464610 - 98546380 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01306722_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ikzf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011771.1 NP_035901.1 3 - 4 447 77
GenBank mRNA AF001293.1 - 2 - 3 166 77
AI549961.1 - 3 - 4 193 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003680.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
regulation of B cell proliferation
B cell activation
regulation of apoptotic process
regulation of B cell differentiation
regulation of lymphocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
nucleic acid binding
DNA binding
protein binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100033889.mm.1 NM_011771.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033889.mm.1 NM_011771.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100033889.mm.1 TC1100003680.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0