This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01307131_g1
See other CYB5R3 GE Assays ›
Gene Symbol:
Cyb5r3
Gene Name:
cytochrome b5 reductase 3
Gene Aliases:
0610016L08Rik, 2500002N19Rik, C85115, Dia-1, Dia1, WU:AL591952.1-001, WU:AL591952.1-002, WU:AL591952.1-003, WU:Cyb5r3
Chromosome Location:
Chr.15: 83153495 - 83172208 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01307131_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cyb5r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029787.2 NP_084063.1 6 - 7 623 63
GenBank mRNA AF332059.1 - 6 - 7 547 63
AF332060.1 - 6 - 7 547 63
AK002640.1 - 6 - 7 595 63
AK010858.1 - 6 - 7 600 63
AK128917.1 - 6 - 7 617 63
AK181262.1 - 2 - 3 121 63
AK183532.1 - 2 - 3 121 63
AK190014.1 - 2 - 3 121 63
AK211940.1 - 2 - 3 121 63
AK211950.1 - 2 - 3 121 63
AK213922.1 - 2 - 3 121 63
BC004760.1 - 6 - 7 630 3154531 63
BC032013.1 - 6 - 7 605 4238275 63
BC043074.1 - 6 - 7 623 6404746 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001967.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cyb5r3

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
steroid biosynthetic process
cholesterol biosynthetic process
steroid metabolic process
cholesterol metabolic process
sterol biosynthetic process
oxidation-reduction process
cytochrome-b5 reductase activity, acting on NAD(P)H
protein binding
AMP binding
oxidoreductase activity
ADP binding
flavin adenine dinucleotide binding
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500015508.mm.1 NM_029787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015508.mm.1 NM_029787.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500015508.mm.1 TC1500001967.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500015507.mm.1 NM_029787.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500015507.mm.1 NM_029787.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500015507.mm.1 TC1500001967.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0