This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308034_g1
See other LMF2 GE Assays ›
Gene Symbol:
Lmf2
Gene Name:
lipase maturation factor 2
Gene Aliases:
AI451006, Tmem112b, Tmem153
Chromosome Location:
Chr.15: 89351004 - 89355659 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308034_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lmf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178919.4 NP_849250.1 5 - 6 981 91
GenBank mRNA AK042427.1 - 5 - 6 941 91
AK084606.1 - 5 - 6 807 91
AK149916.1 - 5 - 6 933 91
AK149934.1 - 5 - 6 982 91
AK171488.1 - 5 - 6 962 91
AK192939.1 - 1 - 2 73 91
BC026448.1 - 4 - 5 761 91
BC038039.1 - 4 - 5 1483 91
BC058973.1 - 3 - 4 1115 91
BC079532.1 - 5 - 6 877 6403184 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002050.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lmf2

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016473.mm.1 NM_178919.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016473.mm.1 NM_178919.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500016473.mm.1 TC1500002050.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0