This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308117_g1
See other HDAC10 GE Assays ›
Gene Symbol:
Hdac10
Gene Name:
histone deacetylase 10
Gene Aliases:
AW548891, Hd10
Chromosome Location:
Chr.15: 89123303 - 89128700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308117_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199198.2 NP_954668.2 18 - 19 2083 61
NR_028447.1 18 - 19 2094 61
NR_028448.1 16 - 17 2292 61
NR_028449.1 17 - 18 2372 61
GenBank mRNA AI448228.1 - 2 - 3 61
AK040235.1 - 17 - 18 2324 61
AK045028.1 - 16 - 17 2572 61
AK154045.1 - 16 - 17 2256 61
BC013700.1 - 18 - 19 2047 61
BC064018.1 - 18 - 19 2010 3980699 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002037.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac10

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
chromatin modification
histone deacetylation
negative regulation of transcription, DNA-templated
histone deacetylase activity
hydrolase activity
enzyme binding
histone deacetylase activity (H3-K14 specific)
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
histone deacetylase activity (H4-K16 specific)
histone deacetylase binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
NAD-dependent histone deacetylase activity (H3-K18 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NM_199198.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NM_199198.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028447.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028447.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028448.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028448.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028449.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 NR_028449.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016211.mm.1 TC1500002037.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NM_199198.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NM_199198.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028447.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028447.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028448.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028448.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028449.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 NR_028449.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016212.mm.1 TC1500002037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0