This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308123_g1
See other HDAC10 GE Assays ›
Gene Symbol:
Hdac10
Gene Name:
histone deacetylase 10
Gene Aliases:
AW548891, Hd10
Chromosome Location:
Chr.15: 89123303 - 89128700 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308123_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199198.2 NP_954668.2 5 - 6 745 62
NR_028447.1 5 - 6 745 62
NR_028449.1 5 - 6 745 62
GenBank mRNA AK040235.1 - 5 - 6 697 62
AK045028.1 - 4 - 5 956 62
BC013700.1 - 5 - 6 698 62
BC064018.1 - 5 - 6 672 3980699 62
BU706947.1 - 5 - 6 486 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002037.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac10

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
protein deacetylation
chromatin modification
histone deacetylation
negative regulation of transcription, DNA-templated
histone deacetylase activity
hydrolase activity
enzyme binding
histone deacetylase activity (H3-K14 specific)
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein deacetylase activity
histone deacetylase activity (H4-K16 specific)
histone deacetylase binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
NAD-dependent histone deacetylase activity (H3-K18 specific)

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NM_199198.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NM_199198.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NR_028447.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NR_028447.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NR_028449.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 NR_028449.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016235.mm.1 TC1500002037.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline