This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308166_g1
See other MIOX GE Assays ›
Gene Symbol:
Miox
Gene Name:
myo-inositol oxygenase
Gene Aliases:
0610009I10Rik, AI314022, Aldrl6, C85427, RSOR
Chromosome Location:
Chr.15: 89334473 - 89337007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308166_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Miox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019977.2 NP_064361.2 2 - 3 133 72
GenBank mRNA AF197127.1 - 2 - 3 131 72
AK002406.1 - 2 - 3 147 72
AY738257.1 - 1 - 2 103 72
BC013543.1 - 2 - 3 133 4242159 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000870.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Miox

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

inositol catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
iron ion binding
ferric iron binding
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
inositol oxygenase activity
NADP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500006836.mm.1 NM_019977.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006836.mm.1 NM_019977.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006836.mm.1 TC1500000870.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0