This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308169_g1
See other MIOX GE Assays ›
Gene Symbol:
Miox
Gene Name:
myo-inositol oxygenase
Gene Aliases:
0610009I10Rik, AI314022, Aldrl6, C85427, RSOR
Chromosome Location:
Chr.15: 89334473 - 89337007 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308169_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Miox
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019977.2 NP_064361.2 9 - 10 774 133
GenBank mRNA AF197127.1 - 9 - 10 772 133
AK002406.1 - 9 - 10 787 133
AY738257.1 - 8 - 9 744 133
BC013543.1 - 9 - 10 774 4242159 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000870.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Miox

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

inositol catabolic process
oxidation-reduction process
aldo-keto reductase (NADP) activity
iron ion binding
ferric iron binding
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
metal ion binding
inositol oxygenase activity
NADP binding

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023200 BC013543.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023200 AK165481.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023200 AK002406.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023200 NM_019977.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84166 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023200 BC013543.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023200 AK165481.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023200 AK002406.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023200 NM_019977.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P385258 AF197127.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424608 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424626 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424607 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424627 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424617 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424609 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424579 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424577 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424612 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424623 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424622 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424628 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424621 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424618 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424602 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424613 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424574 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424625 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424600 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424603 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424619 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424605 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424624 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424611 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424614 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424597 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30424616 - -
Whole Mouse Genome G4122A A_51_P385258 AF197127.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109636 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12296047 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12296051 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12296045 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424637 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424616 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424602 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424590 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424627 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424611 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424593 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424619 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424574 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424622 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30424655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12016077 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424574 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424597 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424619 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424607 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424593 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424633 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12260179 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424591 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424603 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12423565 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424596 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424595 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12404249 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12260171 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12450434 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424600 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424592 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12369267 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12422823 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424638 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424611 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424598 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424637 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424584 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12296050 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424579 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424602 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424583 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424608 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12296048 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P11946384 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12296021 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424624 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424601 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424590 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424609 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424612 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424613 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424605 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12296043 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30424641 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10426218 - -
Affymetrix Mouse Gene 1.0 ST Array 10426220 - -
Affymetrix Mouse Gene 1.0 ST Array 10426215 - -
Affymetrix Mouse Gene 1.0 ST Array 10426222 - -
Affymetrix Mouse Gene 1.0 ST Array 10426223 - -
Affymetrix Mouse Gene 1.0 ST Array 10426216 - -
Affymetrix Mouse Gene 1.0 ST Array 10426214 - -
Affymetrix Mouse Gene 1.0 ST Array 10426219 - -
Affymetrix Mouse Gene 1.0 ST Array 10426224 - -
Affymetrix Mouse Gene 1.0 ST Array 10426221 - -
Affymetrix HT MG-430 PM Array Plate 1416460_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10426213 - -
Mouse Genome 430 2.0 Array 1416460_at - -
Mouse Genome 430 2.0 Array 1416460_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 56727_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 56727_PM_at - -
Affymetrix Mouse Clariom S Array PSR1500006850.mm.1 NM_019977.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500006850.mm.1 NM_019977.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500006850.mm.1 TC1500000870.mm.1 MTA TC
Mouse Expression Set 430 1416460_PM_at - -
Mouse Expression Set 430 1416460_at - -
Murine Genome U74v2 Set 93781_PM_at - -
Murine Genome U74v2 Set 93781_at - -
Mouse Genome 430A 2.0 Array 1416460_PM_at - -
Mouse Genome 430A 2.0 Array 1416460_at - -
Affymetrix HT Mouse Genome 430A Array 1416460_at - -
Affymetrix HT Mouse Genome 430A Array 1416460_PM_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 4810364 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004810364 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2640116 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2640116 - -
mouse-6 v1.1 (Illumina) scl47618.10.1_16-S - -
Illumina Sentrix mouse-6.v1 bead array 4810364 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl47618.10.1_16-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810364 - -
MouseRef-8 v2 Expression BeadChip ILMN_2640116 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2640116 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0