This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308354_m1
See other GRASP GE Assays ›
Gene Symbol:
Grasp
Gene Name:
GRP1 (general receptor for phosphoinositides 1)-associated scaffold protein
Gene Aliases:
tamalin
Chromosome Location:
Chr.15: 101224207 - 101232756 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308354_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grasp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019518.3 NP_062391.3 4 - 5 546 97
GenBank mRNA AB041603.1 - 4 - 5 544 97
AF236099.1 - 4 - 5 509 97
BC046307.1 - 4 - 5 514 6306542 97
BY730445.1 - 4 - 5 565 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001065.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grasp

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
signal transduction
protein localization
protein binding
PDZ domain binding
ADP-ribosylation factor binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008479.mm.1 NM_019518.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008479.mm.1 NM_019518.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500008479.mm.1 TC1500001065.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0