This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308531_m1
See other IGSF23 GE Assays ›
Gene Symbol:
Igsf23
Gene Name:
immunoglobulin superfamily, member 23
Gene Aliases:
2210010C17Rik
Chromosome Location:
Chr.7: 19937305 - 19950765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308531_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027308.2 NP_081584.1 1 - 2 301 93
GenBank mRNA AK008696.1 - 1 - 2 303 93
AK033677.1 - 1 - 2 302 93
BC139455.1 - 1 - 2 149 8862477 93
BC139458.1 - 1 - 2 149 8862480 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 319444 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0