This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01308532_m1
See other IGSF23 GE Assays ›
Gene Symbol:
Igsf23
Gene Name:
immunoglobulin superfamily, member 23
Gene Aliases:
2210010C17Rik
Chromosome Location:
Chr.7: 19937305 - 19950765 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01308532_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027308.2 NP_081584.1 2 - 3 508 68
GenBank mRNA AK008696.1 - 2 - 3 510 68
AK033677.1 - 2 - 3 509 68
AK088927.1 - 3 - 4 68
AK135657.1 - 314 68
BC139455.1 - 2 - 3 356 8862477 68
BC139458.1 - 2 - 3 356 8862480 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002509.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022223.mm.1 TC0700002509.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022223.mm.1 NM_027308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022223.mm.1 NM_027308.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022220.mm.1 TC0700002509.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022220.mm.1 NM_027308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022220.mm.1 NM_027308.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022221.mm.1 TC0700002509.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022221.mm.1 NM_027308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022221.mm.1 NM_027308.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022224.mm.1 TC0700002509.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022224.mm.1 NM_027308.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022224.mm.1 NM_027308.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0